(3-Fluoro-5-iodo-2-isopropoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

57993-500MG 57993-1G 57993-5G 57993-25G
77330-080EA 149.35 USD
77330-080 77345-310 77366-374 77388-094
(3-Fluoro-5-iodo-2-isopropoxyphenyl)methanol ≥97%
(3-Fluoro-5-iodo-2-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂FIO₂
MW: 310.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758413
CAS Number: 2807470-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR