5-Bromo-1-fluoro-2-isopropoxy-3-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

57984-500MG 57984-1G 57984-5G
77340-240EA 854.9 USD
77340-240 77354-082 77378-288
5-Bromo-1-fluoro-2-isopropoxy-3-(methoxymethyl)benzene ≥97%
5-Bromo-1-fluoro-2-isopropoxy-3-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrFO₂
MW: 277.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758405
CAS Number: 2764728-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR