(2-(Benzyloxy)-5-bromo-3-fluorophenyl)methanol ≥97%
Supplier: AOBChem USA
|
Warning
|
57947-500MG
57947-1G
57947-5G
57947-25G
77330-068EA
231.75
USD
77330-068
77354-062
77366-362
77388-082
(2-(Benzyloxy)-5-bromo-3-fluorophenyl)methanol ≥97%
(2-(Benzyloxy)-5-bromo-3-fluorophenyl)methanol
Precaución: for Research Use Only
Formula:
C₁₄H₁₂BrFO₂ MW: 311.146 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28063074 CAS Number: 1824017-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...