(5-Bromo-3-fluoro-2-isopropoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

57945-500MG 57945-1G 57945-5G
77330-066EA 315.18 USD
77330-066 77354-060 77378-272
(5-Bromo-3-fluoro-2-isopropoxyphenyl)methanol ≥97%
(5-Bromo-3-fluoro-2-isopropoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
MW: 263.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758375
CAS Number: 2156117-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR