(5-Bromo-2-ethoxy-3-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

57942-500MG 57942-1G 57942-5G
77330-064EA 337.84 USD
77330-064 77354-056 77378-268
(5-Bromo-2-ethoxy-3-fluorophenyl)methanol ≥97%
(5-Bromo-2-ethoxy-3-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
MW: 249.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28063051
CAS Number: 1823949-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR