5-Bromo-2-ethoxy-3-fluorobenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
57941-500MG
57941-1G
57941-25G
57941-5G
77330-062EA
306.94
USD
77330-062
77354-054
77388-080
77378-266
5-Bromo-2-ethoxy-3-fluorobenzaldehyde ≥97%
5-Bromo-2-ethoxy-3-fluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₉H₈BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD28063047 CAS Number: 1823336-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...