3-Fluoro-5-iodo-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57940-500MG 57940-1G 57940-5G
77338-454EA 489.25 USD
77338-454 77354-052 77378-264
3-Fluoro-5-iodo-2-methoxybenzaldehyde ≥97%
3-Fluoro-5-iodo-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
Storage Temperature: Ambient
MDL Number: MFCD29923370
CAS Number: 1956367-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR