(3-Bromo-2-chloro-5-ethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

57878-500MG 57878-1G 57878-5G
77338-768EA 517.06 USD
77338-768 77354-050 77378-260
(3-Bromo-2-chloro-5-ethylphenyl)methanol ≥97%
(3-Bromo-2-chloro-5-ethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758331
CAS Number: 2710854-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR