3-Bromo-2-chloro-5-ethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

57875-500MG 57875-1G 57875-5G
77338-260EA 470.71 USD
77338-260 77354-048 77378-258
3-Bromo-2-chloro-5-ethylbenzoic acid ≥95%
3-Bromo-2-chloro-5-ethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD28505591
CAS Number: 1935296-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR