1,2,3-Tribromo-5-ethylbenzene ≥97%

Supplier: AOBChem USA

Warning

57872-500MG 57872-1G 57872-5G 57872-25G
77330-058EA 120.51 USD
77330-058 77343-754 77366-356 77378-252
1,2,3-Tribromo-5-ethylbenzene ≥97%
1,2,3-Tribromo-5-ethylbenzene

Precaución: for Research Use Only

Formula: C₈H₇Br₃
MW: 342.853 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758328
CAS Number: 2310221-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR