3-Fluoro-N-methoxy-N,2-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

57864-500MG 57864-1G 57864-5G 57864-25G
77330-056EA 179.22 USD
77330-056 77346-728 77366-354 77388-076
3-Fluoro-N-methoxy-N,2-dimethylbenzamide ≥97%
3-Fluoro-N-methoxy-N,2-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO₂
Storage Temperature: Ambient
MDL Number: MFCD34564503
CAS Number: 1934698-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR