(4,5-Difluoro-2-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

57857-500MG 57857-1G 57857-5G
77336-100EA 375.95 USD
77336-100 77354-038 77378-248
(4,5-Difluoro-2-iodophenyl)methanol ≥97%
(4,5-Difluoro-2-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅F₂IO
Storage Temperature: Ambient
MDL Number: MFCD32705630
CAS Number: 2091032-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR