4,5-Difluoro-2-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57856-500MG 57856-1G 57856-5G
77330-054EA 336.81 USD
77330-054 77354-036 77378-246
4,5-Difluoro-2-iodobenzaldehyde ≥97%
4,5-Difluoro-2-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃F₂IO
MW: 267.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28789188
CAS Number: 1807085-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR