4,5-Difluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57855-500MG 57855-1G 57855-5G 57855-25G
77330-052EA 131.84 USD
77330-052 77344-372 77366-352 77388-074
4,5-Difluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97%
4,5-Difluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₂O₃
MW: 268.064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758325
CAS Number: 2776201-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR