1-Fluoro-5-iodo-2-isopropoxy-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

57854-500MG 57854-1G 57854-5G 57854-25G
77330-050EA 187.46 USD
77330-050 77354-034 77366-350 77388-072
1-Fluoro-5-iodo-2-isopropoxy-3-(trifluoromethyl)benzene ≥97%
1-Fluoro-5-iodo-2-isopropoxy-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₉F₄IO
MW: 348.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758324
CAS Number: 2764728-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR