2-Ethoxy-1-fluoro-5-iodo-3-(trifluoromethyl)benzene ≥95%
Supplier: AOBChem USA
|
Warning
|
57852-500MG
57852-1G
57852-5G
57852-25G
77330-046EA
179.22
USD
77330-046
77346-726
77366-346
77388-068
2-Ethoxy-1-fluoro-5-iodo-3-(trifluoromethyl)benzene ≥95%
2-Ethoxy-1-fluoro-5-iodo-3-(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₉H₇F₄IO Storage Temperature: Ambient |
MDL Number:
MFCD34758322 CAS Number: 2386475-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...