3-(3-Bromophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%

Supplier: AOBChem USA

Warning

57813-500MG 57813-1G 57813-5G 57813-25G
77330-040EA 99.91 USD
77330-040 77342-838 77366-344 77378-240
3-(3-Bromophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%
3-(3-Bromophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO₃
MW: 288.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758308
CAS Number: 2807446-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR