5-Chloro-N-methoxy-2-(trifluoromethyl)benzamide ≥95%

Supplier: AOBChem USA

Warning

57803-500MG 57803-1G 57803-5G
77330-038EA 319.3 USD
77330-038 77354-028 77378-234
5-Chloro-N-methoxy-2-(trifluoromethyl)benzamide ≥95%
5-Chloro-N-methoxy-2-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₉H₇ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD34530618
CAS Number: 2734774-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR