4-Fluoro-2-isopropoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

57791-500MG 57791-1G 57791-5G
77340-316EA 900.22 USD
77340-316 77354-026 77378-232
4-Fluoro-2-isopropoxy-5-methylbenzoic acid ≥97%
4-Fluoro-2-isopropoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758302
CAS Number: 2764731-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR