2-(Benzyloxy)-4-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

57775-500MG 57775-1G 57775-5G
77335-418EA 353.29 USD
77335-418 77354-020 77378-226
2-(Benzyloxy)-4-fluoro-5-methylbenzoic acid ≥97%
2-(Benzyloxy)-4-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564489
CAS Number: 2734774-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR