1-Ethoxy-5-fluoro-2-iodo-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

57773-500MG 57773-1G 57773-5G 57773-25G
77330-036EA 254.41 USD
77330-036 77354-018 77366-338 77388-066
1-Ethoxy-5-fluoro-2-iodo-4-methylbenzene ≥97%
1-Ethoxy-5-fluoro-2-iodo-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO
MW: 280.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564487
CAS Number: 2091046-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR