(2-Ethoxy-4-fluoro-5-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

57772-500MG 57772-1G 57772-5G
77340-314EA 900.22 USD
77340-314 77354-016 77378-224
(2-Ethoxy-4-fluoro-5-methylphenyl)methanol ≥95%
(2-Ethoxy-4-fluoro-5-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34564486
CAS Number: 2090511-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR