Methyl 2-ethoxy-4-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

57770-500MG 57770-1G 57770-5G
77335-822EA 366.68 USD
77335-822 77354-014 77378-222
Methyl 2-ethoxy-4-fluoro-5-methylbenzoate ≥97%
Methyl 2-ethoxy-4-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD34758293
CAS Number: 2383042-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR