1-Cyclopentyl-4-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

57767-500MG 57767-1G 57767-5G
77340-076EA 746.75 USD
77340-076 77354-010 77378-216
1-Cyclopentyl-4-iodobenzene ≥95%
1-Cyclopentyl-4-iodobenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₃I
Storage Temperature: Ambient
MDL Number: MFCD11111749
CAS Number: 92316-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR