Ethyl 4-cyclopentylbenzoate ≥95%

Supplier: AOBChem USA

Warning

57765-500MG 57765-1G 57765-5G
77339-372EA 582.98 USD
77339-372 77354-008 77378-214
Ethyl 4-cyclopentylbenzoate ≥95%
Ethyl 4-cyclopentylbenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34758292
CAS Number: 1448351-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR