(2-Ethoxy-5-ethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

57755-500MG 57755-1G 57755-5G 57755-25G
77330-034EA 202.91 USD
77330-034 77354-006 77366-334 77388-064
(2-Ethoxy-5-ethylphenyl)methanol ≥97%
(2-Ethoxy-5-ethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD34535611
CAS Number: 1528483-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR