3-Bromo-5-fluoro-N,2-dimethoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

57750-500MG 57750-1G 57750-5G 57750-25G
77330-030EA 226.6 USD
77330-030 77354-004 77366-330 77388-060
3-Bromo-5-fluoro-N,2-dimethoxy-N-methylbenzamide ≥97%
3-Bromo-5-fluoro-N,2-dimethoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrFNO₃
MW: 292.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758289
CAS Number: 2755724-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR