(2,4-Dibromo-6-fluorophenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

57731-500MG 57731-1G 57731-5G 57731-25G
77330-020EA 202.91 USD
77330-020 77353-994 77366-320 77388-050
(2,4-Dibromo-6-fluorophenyl)(thiomorpholino)methanone ≥97%
(2,4-Dibromo-6-fluorophenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀Br₂FNOS
MW: 383.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758275
CAS Number: 2755718-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR