(2,4-Dibromo-6-fluorophenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

57730-500MG 57730-1G 57730-5G 57730-25G
77330-018EA 202.91 USD
77330-018 77353-992 77366-318 77388-048
(2,4-Dibromo-6-fluorophenyl)(morpholino)methanone ≥97%
(2,4-Dibromo-6-fluorophenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀Br₂FNO₂
MW: 367.009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758274
CAS Number: 2755723-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR