(2,4-Dibromo-6-fluorophenyl)(pyrrolidin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
57729-500MG
57729-1G
57729-5G
57729-25G
77330-016EA
206
USD
77330-016
77353-990
77366-316
77388-046
(2,4-Dibromo-6-fluorophenyl)(pyrrolidin-1-yl)methanone ≥97%
(2,4-Dibromo-6-fluorophenyl)(pyrrolidin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₁₁H₁₀Br₂FNO MW: 351.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34758273 CAS Number: 2764728-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...