2,4-Dibromo-N-cyclohexyl-6-fluorobenzamide ≥95%

Supplier: AOBChem USA

Warning

57728-500MG 57728-1G 57728-5G 57728-25G
77330-014EA 189.52 USD
77330-014 77353-988 77366-314 77388-044
2,4-Dibromo-N-cyclohexyl-6-fluorobenzamide ≥95%
2,4-Dibromo-N-cyclohexyl-6-fluorobenzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₄Br₂FNO
Storage Temperature: Ambient
MDL Number: MFCD34758272
CAS Number: 2755719-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR