2,4-Dibromo-N-cyclopropyl-6-fluorobenzamide ≥95%

Supplier: AOBChem USA

Warning

57727-500MG 57727-1G 57727-5G 57727-25G
77330-012EA 187.46 USD
77330-012 77353-986 77366-312 77388-042
2,4-Dibromo-N-cyclopropyl-6-fluorobenzamide ≥95%
2,4-Dibromo-N-cyclopropyl-6-fluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₈Br₂FNO
Storage Temperature: Ambient
MDL Number: MFCD34758271
CAS Number: 2755721-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR