6-Chloro-2,4'-difluoro-3-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

57725-500MG 57725-1G 57725-5G
77337-450EA 414.06 USD
77337-450 77353-984 77378-210
6-Chloro-2,4'-difluoro-3-methyl-1,1'-biphenyl ≥97%
6-Chloro-2,4'-difluoro-3-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉ClF₂
MW: 238.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758269
CAS Number: 2755722-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR