5-Bromo-3-(tert-butyl)-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

57705-500MG 57705-1G 57705-5G
77336-708EA 392.43 USD
77336-708 77353-980 77378-206
5-Bromo-3-(tert-butyl)-2-methoxybenzoic acid ≥97%
5-Bromo-3-(tert-butyl)-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₃
MW: 287.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758263
CAS Number: 1897098-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR