5-Bromo-3-(tert-butyl)-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57701-500MG 57701-1G 57701-5G
77330-008EA 318.27 USD
77330-008 77353-978 77378-204
5-Bromo-3-(tert-butyl)-2-methoxybenzaldehyde ≥97%
5-Bromo-3-(tert-butyl)-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₅BrO₂
Storage Temperature: Ambient
MDL Number: MFCD22571441
CAS Number: 417715-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR