3-Chloro-4-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

57700-500MG 57700-1G 57700-5G
77336-634EA 390.37 USD
77336-634 77353-976 77378-202
3-Chloro-4-fluoro-1,1'-biphenyl ≥97%
3-Chloro-4-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈ClF
MW: 206.643 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758259
CAS Number: 1803002-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR