(3'-Chloro-4'-fluoro-2-methyl-[1,1'-biphenyl]-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

57699-500MG 57699-1G 57699-5G
77339-596EA 602.55 USD
77339-596 77353-974 77378-200
(3'-Chloro-4'-fluoro-2-methyl-[1,1'-biphenyl]-4-yl)methanol ≥97%
(3'-Chloro-4'-fluoro-2-methyl-[1,1'-biphenyl]-4-yl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClFO
MW: 250.696 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758258
CAS Number: 2755721-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR