2,4-Dibromo-6-fluoro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

57696-500MG 57696-1G 57696-5G
77337-448EA 414.06 USD
77337-448 77353-972 77378-198
2,4-Dibromo-6-fluoro-N-methylbenzamide ≥97%
2,4-Dibromo-6-fluoro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₆Br₂FNO
Storage Temperature: Ambient
MDL Number: MFCD34758257
CAS Number: 2755723-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR