5-Bromo-[1,1'-biphenyl]-2-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

57693-500MG 57693-1G 57693-5G
77337-518EA 416.12 USD
77337-518 77353-970 77378-196
5-Bromo-[1,1'-biphenyl]-2-carbaldehyde ≥95%
5-Bromo-[1,1'-biphenyl]-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD28061558
CAS Number: 1675969-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR