4,4'-Difluoro-3-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

57671-500MG 57671-1G 57671-5G
77330-000EA 321.36 USD
77330-000 77353-962 77378-188
4,4'-Difluoro-3-methoxy-1,1'-biphenyl ≥97%
4,4'-Difluoro-3-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂O
MW: 220.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14699123
CAS Number: 479190-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR