4-Fluoro-3-methoxy-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

57668-500MG 57668-1G 57668-5G
77336-882EA 396.55 USD
77336-882 77353-960 77378-186
4-Fluoro-3-methoxy-4'-methyl-1,1'-biphenyl ≥97%
4-Fluoro-3-methoxy-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃FO
MW: 216.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758251
CAS Number: 2755724-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR