2-Bromo-4-(tert-butyl)-4'-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

57665-500MG 57665-1G 57665-5G
77335-570EA 357.41 USD
77335-570 77353-958 77378-184
2-Bromo-4-(tert-butyl)-4'-fluoro-1,1'-biphenyl ≥97%
2-Bromo-4-(tert-butyl)-4'-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₆BrF
MW: 307.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564479
CAS Number: 2734779-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR