4-Fluoro-3-methoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

57651-500MG 57651-1G 57651-5G
77336-880EA 396.55 USD
77336-880 77353-956 77378-182
4-Fluoro-3-methoxy-1,1'-biphenyl ≥97%
4-Fluoro-3-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁FO
MW: 202.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701858
CAS Number: 1092656-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR