1-(Benzyloxy)-2-bromo-4-ethylbenzene ≥97%

Supplier: AOBChem USA

Warning

57617-1G 57617-5G 57617-25G
77341-346EA 82.4 USD
77341-346 77366-300 77378-178
1-(Benzyloxy)-2-bromo-4-ethylbenzene ≥97%
1-(Benzyloxy)-2-bromo-4-ethylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO
MW: 291.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00838271
CAS Number: 127971-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR