2-Bromo-4-ethyl-1-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

57616-1G 57616-5G 57616-25G
77341-258EA 79.31 USD
77341-258 77366-298 77378-176
2-Bromo-4-ethyl-1-isopropoxybenzene ≥97%
2-Bromo-4-ethyl-1-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356527
CAS Number: 1369878-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR