2-Ethoxy-5-ethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57615-500MG 57615-1G 57615-5G 57615-25G
77329-996EA 107.12 USD
77329-996 77343-130 77366-296 77378-174
2-Ethoxy-5-ethylbenzaldehyde ≥97%
2-Ethoxy-5-ethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06247338
CAS Number: 883536-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR