Ethyl 2-ethoxy-5-ethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

57614-500MG 57614-1G 57614-5G 57614-25G
77329-994EA 259.56 USD
77329-994 77353-952 77366-294 77388-030
Ethyl 2-ethoxy-5-ethylbenzoate ≥95%
Ethyl 2-ethoxy-5-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31773588
CAS Number: 1369368-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR