Methyl 5-ethyl-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

57605-500MG 57605-1G 57605-5G
77329-990EA 332.69 USD
77329-990 77353-948 77378-168
Methyl 5-ethyl-2-methoxybenzoate ≥95%
Methyl 5-ethyl-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD17167548
CAS Number: 66873-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR