2,4-Dichloro-3-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

57579-500MG 57579-1G 57579-5G
77329-988EA 309 USD
77329-988 77353-944 77378-164
2,4-Dichloro-3-methylbenzamide ≥97%
2,4-Dichloro-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD28786757
CAS Number: 1806303-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR