4'-Bromo-2',5'-difluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

57560-500MG 57560-1G 57560-5G 57560-25G
77329-982EA 229.69 USD
77329-982 77353-940 77366-284 77388-024
4'-Bromo-2',5'-difluoro-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Bromo-2',5'-difluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₂O
MW: 297.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564458
CAS Number: 2734777-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR